Learn More
ε-Caprolactone, Sigma-Aldrich™
Specifications
Specifications
| CAS | 502-44-3 |
| Melting Point | -1.5°C |
| Density | 1.07 g/cm3 at 20°C |
| Boiling Point | 235°C/1013 hPa |
| MDL Number | MFCD00003267 |
| Quantity | 250 mL |
| Synonym | ε-Caprolactone, 2-Oxepanone, 6-Hydroxycaproic acid lactone, ε-Caprolactone |
| Molecular Weight (g/mol) | 114.14 |
| Grade | Research |