Learn More
Capmatinib, MedChemExpress
Product Code. 16491827
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491827 50 mg 50mg
16478357 5 mg 5mg
16471076 200 mg 200mg
16466176 100 mg 100mg
16425668 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16491827 Supplier Medchem Express Supplier No. HY13404
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
MedChemExpress Capmatinib (INC280; INCB28060) is a potent, orally active, selective, and ATP competitive c-Met kinase inhibitor (IC50=0.13 nM).
Capmatinib can inhibit phosphorylation of c-MET as well as c-MET pathway downstream effectors such as ERK1/2, AKT, FAK, GAB1, and STAT3/5. Capmatinib potently inhibits c-MET-dependent tumor cell proliferation and migration and effectively induces apoptosis. Antitumor activity. Capmatinib is largely metabolized by CYP3A4 and aldehyde oxidase.
Specifications
Chemical Name or Material Capmatinib
CAS 1029712-80-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H17FN6O
Quantity 50 mg
Synonym INC280 INCB28060
Solubility Information DMSO : 25 mg/mL (60.62 mM; Need ultrasonic) ∣H2O : 10 mg/mL (24.25 mM; Need ultrasonic)
SMILES O=C(NC)C1=CC=C(C2=NN3C(N=C2)=NC=C3CC4=CC=C5N=CC=CC5=C4)C=C1F
Molecular Weight (g/mol) 412.42
Formula Weight 412.42
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease