Learn More
Thermo Scientific™ Capitol Vial 16 x 93mm Flat Bottom Sample Tubes
GREENER_CHOICE
Product Code. 17155301
Click to view available options
Unit Size:
Pack of 1000
Product Code. 17155301
Brand: Thermo Scientific™ S40120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Capitol Vial 16 x 93mm Flat Bottom Sample Tubes are ideal for sample collection, storage and transport.
Thermo Scientific™ Capitol Vial 16 x 93mm Flat Bottom Sample Tubes are ideal for sample collection, storage and transport.
  • Compatible with universal plug cap part number M995133
  • Tubes are constrcucted from polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Description 16 x 93mm Flat bottom transfer tubes; Case of 1000
Diameter (Metric) Inner 16 mm
Material Polypropylene
Volume (Metric) 18 mL
No. per Case 1000
Style Flat Bottom
Closure Type Plug
Height (Metric) 93 mm
Certifications/Compliance Certificate of Analysis (COA)
Type Flat Bottom Sample Tube
For Use With (Equipment) Universal Plug Cap - M995133