Learn More
- Home
- Products
- Tubes
- Capillary Tubes
- Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Product Code. 10409712
Change view
Click to view available options
Length (Metric) Tube:
60 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Inner:
0.80 mm
0.95 mm
1.15 mm
1.30 mm
1.55 mm
Unit Size:
Pack of 1000
21 product options available for selection
| Product Code. | Length (Metric) Tube | Diameter (Metric) Inner | unitSize |
|---|---|---|---|
| 10409712 | 80 mm | 1.55 mm | Pack of 1000 |
| 10769191 | 70 mm | 0.80 mm | Pack of 1000 |
| 10673192 | 75 mm | 0.80 mm | Pack of 1000 |
| 10033961 | 80 mm | 0.80 mm | Pack of 1000 |
| 10355511 | 90 mm | 0.80 mm | Pack of 1000 |
| 10253861 | 120 mm | 0.80 mm | Pack of 1000 |
| 10720233 | 60 mm | 0.95 mm | Pack of 1000 |
| 10223531 | 90 mm | 0.95 mm | Pack of 1000 |
| 10006854 | 100 mm | 0.95 mm | Pack of 1000 |
| 10380182 | 120 mm | 0.95 mm | Pack of 1000 |
| 10009760 | 70 mm | 1.15 mm | Pack of 1000 |
| 10613202 | 120 mm | 1.15 mm | Pack of 1000 |
| 10029770 | 150 mm | 1.15 mm | Pack of 1000 |
| 10089650 | 70 mm | 1.30 mm | Pack of 1000 |
| 10673002 | 75 mm | 1.30 mm | Pack of 1000 |
| 10009750 | 100 mm | 1.30 mm | Pack of 1000 |
| 10161721 | 70 mm | 1.55 mm | Pack of 1000 |
| 10223341 | 75 mm | 1.55 mm | Pack of 1000 |
| 10089730 | 90 mm | 1.55 mm | Pack of 1000 |
| 10233671 | 100 mm | 1.55 mm | Pack of 1000 |
| 10059720 | 120 mm | 1.55 mm | Pack of 1000 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Capillary tubes
- One end closed
- Available in a wide range of diameters/lengths
Specifications
Specifications
| Product Type | Melting Point Determination Tube, One End Closed |
| Diameter (Metric) Inner | 1.55 mm |
| Length (Metric) Tube | 80 mm |
| Quantity | 1 x 1000/Pk. |