Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Product Code. 10223531
Click to view available options
Length (Metric) Tube:
60 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Inner:
0.80 mm
0.95 mm
1.15 mm
1.30 mm
1.55 mm
Unit Size:
Pack of 1000
Product Code. 10223531
Brand: Hirschmann™ 9211390
Excellent for melting point analysis in the laboratory
  • Capillary tubes
  • One end closed
  • Available in a wide range of diameters/lengths
Specifications
Product Type Melting Point Determination Tube, One End Closed
Diameter (Metric) Inner 0.95 mm
Length (Metric) Tube 90 mm
Quantity 10 x 100/Pk.