Learn More
Fisherbrand™ Capillary tubes
Product Code. 12328416
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12328416 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12328416 Supplier Fisherbrand™ Supplier No. TV100CAPROD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessory for TV100 standard cooled twin-plate mini-gel electrophoresis unit.
  • Tubes for holding capillary gels
  • Internal diameter: 1mm
  • Used for protein isolation
Specifications
Product Type Capillary Tube
Quantity 10/Pk.
Length (Metric) 1 mm