Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Product Code. 13270490
Change view
Click to view available options
Length (Metric):
10 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Outer:
1 mm
1.35 mm
1.55 mm
1.75 mm
2.35 mm
Unit Size:
Pack of 1000
Product Code. Length (Metric) Diameter (Metric) Outer unitSize
13270490 80 mm 1.35 mm Pack of 1000
10106960 100 mm 1.55 mm Pack of 1000
13240490 10 mm 1 mm Pack of 1000
10489512 150 mm 1.35 mm Pack of 1000
10790033 150 mm 1.75 mm Pack of 1000
10365511 150 mm 1 mm Pack of 1000
10395411 150 mm 2.35 mm Pack of 1000
11807003 80 mm 1.55 mm Pack of 1000
10190851 80 mm 1.75 mm Pack of 1000
10190801 90 mm 1.55 mm Pack of 1000
10652812 90 mm 1.75 mm Pack of 1000
10645214 75 mm 1.35 mm Pack of 1000
10039630 70 mm 1.35 mm Pack of 1000
10587441 75 mm 1.55 mm Pack of 1000
10253671 120 mm 1.75 mm Pack of 1000
15 options
Product Code. 13270490
Brand: Hirschmann™ 9211380
Only null left
Add to basket
Add to basket
Excellent for melting point analysis in the laboratory. Hirschmann Instruments™ Capillary Tubes for Melting Point Determination, One End Closed are manufactured from clear glass.
  • Capillary tubes
  • Available in a range of diameters/lengths
Specifications
Diameter (Metric) Inner 0.95 mm
Diameter (Metric) Outer 1.35 mm
For Use With (Application) Melting point determination
Tube Style One end closed
Length (Metric) 80 mm
Type Capillary Tube