Learn More
IKA™ Capillary Set
Product Code. 17563636 Shop All IKA Products
Change view
Click to view available options
:
Capillary; Includes: Four Different Sizes
Product Code.
17563636 Capillary; Includes: Four Different Sizes
1 options
Product Code. 17563636
Brand: IKA™ 0020103091
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The capillary is used to regulate the flow rate range. It is determined by the size of the capillary and viscosity of the solvent.
  • For regulating the flow rate range.
  • It is determined by the size of the capillary and viscosity of the solvent.
  • The set contains capillary tubes with four different sizes.
  • Scope of delivery
    • Capillary assembly/125 μm/50 mm
    • Capillary assembly/125 μm/100 mm
    • Capillary assembly/180 μm/50 mm
    • Capillary assembly/250 μm/100 mm
TRUSTED_SUSTAINABILITY
Specifications
Includes Four Different Sizes