missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: EVNGVIAGVA PSVEAFDKLM DSMVAEFLKN SRILAGDVET HAEMVHSAFQ AQRAFLLMAS QYQQPHENDV AALLKPISE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.
This gene was identified by its similarity to the gene for human adenylyl cyclase-associated protein. The function of the protein encoded by this gene is unknown. However, the protein appears to be able to interact with adenylyl cyclase-associated protein and actin.
Specifications
Specifications
| Antigen | CAP2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Cap2 |
| Gene Accession No. | P40123 |
| Gene Alias | 2810452G09Rik; Adenylyl cyclase-associated protein 2; AV045235; CAP 2; CAP, adenylate cyclase-associated protein, 2 (yeast); CAP2 |
| Gene Symbols | Cap2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
