Description
- Tightly seals titer plate wells
- Prevents spills and well-to-well contamination
- Made of rugged silicone rubber
- May be sterilized and reused
Specifications
Specifications
| For Use With (Equipment) | 2200 96-Well Titer Plate |
| Material | Silicone Rubber |