Learn More
Canthaxanthine 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30328616
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328616 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328616 Supplier Dr. Ehrenstorfer Supplier No. DREA10947000AL10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Canthaxanthin
CAS 514-78-3
Analyte or Component Names Canthaxanthin
Shipping Condition Room Temperature
Molecular Formula C40 H52 O2
Concentration 10 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym β,β-Carotene-4,4'-dione, β-Carotene-4,4'-dione (6CI), β-Carotene-4,4'-dione, all-trans- (8CI), 4,4'-Diketo-β-carotene, 4,4'-Dioxo-β-carotene, C.I. 40850, C.I. Food Orange 8, Canthaxanthin, Canthaxanthine, Carophyll red, E 161g, Food Orange 8, Lucantin Red, NSC 374110, Roxanthin Red 10, all-trans-Canthaxanthin
InChI Formula InChI=1S/C40H52O2/c1-29(17-13-19-31(3)21-23-35-33(5)37(41)25-27-39(35,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-36-34(6)38(42)26-28-40(36,9)10/h11-24H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C(=O)CCC1(C)C)\C=C\C=C(/C)\C=C\C2=C(C)C(=O)CCC2(C)C
IUPAC Name 2,4,4-trimethyl-3-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaenyl]cyclohex-2-en-1-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 564.84
Quantity 1 mL
Formula Weight 564.3967 g/mol