Learn More
Paragon Scientific™ Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
- Supplied with full UKAS ISO 17025 calibration
Specifications
Specifications
| Viscosity Range | 240 to 1200 cSt |
| Calibrated | Yes |
| Approximate Constant | 1.2 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D445, ISO 3104 and IP 71 Section 1 |
| Includes | UKAS ISO 17025 calibration |
| Size | 400 |
8 week lead-time