missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine Parvovirus Antibody (8H7) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-73204JF669
3871.75 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Canine Parvovirus Monoclonal antibody specifically detects Canine Parvovirus in Virus samples. It is validated for Immunodiffusion,Haemagglutination,Sandwich ELISA,Lateral Flow
Specifications
| Canine Parvovirus | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CPV, Parvo | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine parvovirus. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunodiffusion, Hemagglutination Assay, Sandwich ELISA, Lateral Flow | |
| 8H7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction