Learn More
ACROBiosystems Canine M-CSF R / CSF1R / CD115 Protein, His Tag
Brand:  ACROBiosystems CSR-C82H3-1mg
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16447449
  • 32169.11 NOK / 1mg
Estimated Shipment: 20-01-2023
to see stock.
Description
Description
Colony stimulating factor 1 receptor (CSF1R) is also known as macrophage colony-stimulating factor receptor (M-CSFR), CD115 Cluster of Differentiation 115 (CD115), C-FMS, CSFR, FIM2, FMS, and is a member of the typeIII subfamily of receptor tyrosine kinases (RTKs). CSF1R is a receptor for a cytokine called colony stimulating factor 1, The protein encoded by the CSFR1 gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most, if not all, of the biological effects of this cytokine. Ligand binding activates CSFR1 through a process of oligomerization and transphosphorylation . Mutations in CSF1R are associated with chronic myelomonocytic leukemia and type M4 acute myeloblastic leukemia. Increased levels of CSF1R1 are found in microglia in Alzheimer's disease and after brain injuries. The increased receptor expression causes microglia to become more active. Both CSF1R, and its ligand colony stimulating factor 1 play an important role in the development of the mammary gland and may be involved in the process of mammary gland carcinogenesis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 56.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
XP_546306.2
56.1 kDa
1 mg
Val 20 - Pro 510
CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR
Unconjugated
PBS, pH7.4
HEK293
Canine
Powder
>95% as determined by SDS-PAGE
Canine M-CSF R, His Tag (CSR-C82H3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 510 (Accession # E2RJV2-1).
-20°C
M-CSF R
His Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens