Learn More
Canertinib dihydrochloride, MedChemExpress
Product Code. 16494926
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16494926
Brand: Medchem Express HY10367A
MedChemExpress Canertinib dihydrochloride (CI-1033 dihydrochloride) is a potent and irreversible EGFR inhibitor; inhibits cellular EGFR and ErbB2 autophosphorylation with IC50s of 7.4 and 9 nM. Canertinib dihydrochloride is active against vaccinia virus respiratory infection in mice.
Canertinib dihydrochloride (CI-1033 dihydrochloride) is a potent and irreversible EGFR inhibitor
Specifications
Chemical Name or Material Canertinib dihydrochloride
CAS 289499-45-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27Cl3FN5O3
Quantity 100 mg
Synonym CI-1033 dihydrochloride PD-183805 dihydrochloride
Solubility Information DMSO : 62.5 mg/mL (111.83 mM; Need ultrasonic) ∣H2O : 25 mg/mL (44.73 mM; Need ultrasonic)
SMILES C=CC(NC1=CC2=C(N=CN=C2C=C1OCCCN3CCOCC3)NC4=CC=C(C(Cl)=C4)F)=O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 558.86
Formula Weight 558.86
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.