Learn More
CAMSAP2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17880-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The CAMSAP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CAMSAP2. This antibody recognizes human, mouse, rat antigen. The CAMSAP2 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
CAMSAP2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
23271 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
393 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |