missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech Calretinin Mouse anti-Human, Mouse, Pig, Rat, CoraLite 555, Clone: 2D7A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-66496-100UL
Additional Details : Weight : 0.00010kg
Description
Calretinin Monoclonal antibody apecifically detetects Calretinin antigen in Human, Mouse, Pig, Rat Samples. It is validated for Immunofluorescence.Specifications
Calretinin | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
CALB2 | |
Calretinin fusion protein Ag2924 | |
100 μL | |
794 | |
Store at -20°C. Avoid exposure to light. | |
IgG1 |
Immunofluorescence | |
2D7A9 | |
CoraLite 555 | |
BC015484 | |
Mouse | |
Protein G purification | |
Primary | |
Human, Mouse, Pig, Rat | |
Liquid |