missing translation for 'onlineSavingsMsg'
Learn More

Calponin1 Monoclonal Antibody (OTI3H2), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA813853S

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17508564

  • 4065.15 NOK / 30µL
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

Calponin1 Monoclonal antibody specifically detects Calponin1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Calponin1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P51911, Q08091, Q08290
Mouse
Protein A/G
RUO
1264, 12797, 65204
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin)
OTI3H2
Unconjugated
CNN1
Basic calponin; calponin; calponin 1; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin H1, smooth muscle; Calponin-1; calponins, basic; CN; CNN1; CnnI; epididymis secretory protein Li 14; h1-calponin; HEL-S-14; similar to calponin 1, basic, smooth muscle; Sm-Calp; SMCC; smooth muscle calponin h1
Human recombinant protein fragment corresponding to amino acids 67-257 of human Calponin1 (NP_001290) produced in E.coli.
30 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers