missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin1 Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813852S
Additional Details : Weight : 0.05000kg
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
Calponin1 Monoclonal antibody specifically detects Calponin1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Calponin1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P51911, Q08091, Q08290 | |
Mouse | |
Protein A/G | |
RUO | |
1264, 12797, 65204 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
OTI2D2 | |
Unconjugated | |
CNN1 | |
Basic calponin; calponin; calponin 1; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin H1, smooth muscle; Calponin-1; calponins, basic; CN; CNN1; CnnI; epididymis secretory protein Li 14; h1-calponin; HEL-S-14; similar to calponin 1, basic, smooth muscle; Sm-Calp; SMCC; smooth muscle calponin h1 | |
Human recombinant protein fragment corresponding to amino acids 67-257 of human Calponin1 (NP_001290) produced in E.coli. | |
30 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |