missing translation for 'onlineSavingsMsg'
Learn More

Calponin1 Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene

Product Code. 17558554
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17558554 100 μL 100µL
17578554 30 μL 30µL
2 options
This item is not returnable. View return policy

Product Code. 17558554

Brand: Origene Technologies TA813852

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

Calponin1 Monoclonal antibody specifically detects Calponin1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Calponin1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CNN1
Gene Accession No. P51911, Q08091, Q08290
Gene Alias Basic calponin; calponin; calponin 1; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin H1, smooth muscle; Calponin-1; calponins, basic; CN; CNN1; CnnI; epididymis secretory protein Li 14; h1-calponin; HEL-S-14; similar to calponin 1, basic, smooth muscle; Sm-Calp; SMCC; smooth muscle calponin h1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 67-257 of human Calponin1 (NP_001290) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1264, 12797, 65204
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.