Learn More
Thermo Scientific Chemicals Calpain Inhibitor XI
Product Code. 15407179
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 15407179
Brand: Thermo Scientific Chemicals J65991.#0
Request Bulk or Custom Format
A cell-permeable dipeptidyl --ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145731-49-3
Molecular Formula C26H40N4O6
Synonym Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine
Specifications
Quantity 5 mg
Description Off-white
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable dipeptidyl →-ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2
Formula Weight 504.60
Sensitivity Light Sensitive
Form Solid
InChI Key TZVQRMYLYQNBOA-UHFFFAOYNA-N
SMILES CCC(NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C(=O)C(=O)NCCCN1CCOCC1
IUPAC Name benzyl N-{3-methyl-1-[(1-{[3-(morpholin-4-yl)propyl]carbamoyl}-1-oxobutan-2-yl)carbamoyl]butyl}carbamate
Molecular Weight (g/mol) 504.63