Learn More
CALN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16858583 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16858583 20 μL 20µL
16848583 150 μL 150µL
2 options
Product Code. 16858583
Brand: Proteintech 114771AP20UL
Rabbit Polyclonal Antibody
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP8, contains two EF-hand domains for calcium binding and shares 70% homology with CaBP7 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. Both CaBP8 and CaBP7 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
Specifications
Antigen CALN1
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CALN1
Gene Accession No. Q06BI3, Q9BXU9, Q9JJG7
Gene Alias CaBP8, Calcium binding protein 8, CALN1, calneuron 1, Calneuron I
Gene Symbols Caln1
Host Species Rabbit
Immunogen CALN1 Fusion Protein Ag2043
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140904, 363909, 83698
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG