Learn More
WTW™ Calibration and Maintenance Set
GREENER_CHOICE
Product Code. 10634471
Click to view available options
Type:
Standard Calibration and Maintenance Set
Technical Calibration and Maintenace Set
Unit Size:
1 set
Each
Product Code. 10634471
Brand: WTW™ 109415
Only null left
Add to basket
Add to basket
Calibration and maintenance are complete assortments for the calibration and maintenance of electrodes. The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCI as a storage or refill solution.
  • Adapted to different electrode types
  • Cost-effective combinations
  • Calibration and maintenance in one assortment
TRUSTED_SUSTAINABILITY
Specifications
Type Standard Calibration and Maintenance Set
Includes With standard (DIN/NBS) buffer solution, 3 bottles, 250mL each of pH 4.006 - 6.865 - 9.180, 1 bottle with 250mL pepsin cleaning solution, 1 bottle with 250mL KCl solution 3M/L
compliance-icons
Product Identifier
  • PEP/pH
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • EUH208-Contains Pepsin A. May produce an allergic reaction.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P234-Keep only in original container.
Supplemental information
  • MIXTURE LIST-Contains : pepsin Aborax decahydrate