Learn More
HUBER Calibration Inserts
Click to view available options
For Use With (Equipment):
Ministat 125/w
Ministat 240/w
Unit Size:
Each
Description
Calibration insert Ministat
Specifications
Specifications
| For Use With (Equipment) | Ministat 240/w |
| Volume (Metric) | 4.9 L |
| Quantity | 1 Ea. |
| Weight (Metric) | 1.8 kg |
| Type | Calibration Insert |