Learn More
HUBER Calibration Inserts
Description
Calibration insert Ministat
Specifications
Specifications
| For Use With (Equipment) | Ministat 125/w |
| Volume (Metric) | 2.8 L |
| Quantity | 1 Ea. |
| Weight (Metric) | 1.167 kg |
| Type | Calibration Insert |