Learn More
BINDER™ Calibration Certificate for BINDER™ KB ECO Series
The Calibration Certificate for BINDER KB ECO Series is an option for KB ECO Series Refrigerated Incubators with environmentally friendly Thermoelectric Cooling.
Brand: BINDER™ 8012-2108
Additional Details : Weight : 0.00100kg
Description
- Calibration certificate
- For temperature
Specifications
BINDER Chambers Series KB ECO | |
Calibration certificate, temperature - temperature measurement incl. certificate, 9 measuring points (at specified temperature) | |
Calibration Certificate | |
9 P Temperature |