Learn More
Calhex 231, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Calhex 231 |
| CAS | 652973-93-8 |
| Molecular Formula | C25 H27 Cl N2 O |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C25H27ClN2O/c1-17(21-10-6-8-18-7-2-3-9-22(18)21)27-23-11-4-5-12-24(23)28-25(29)19-13-15-20(26)16-14-19/h2-3,6-10,13-17,23-24,27H,4-5,11-12H2,1H3,(H,28,29)/t17-,23+,24+/m1/s1 |
| SMILES | C[C@@H](N[C@H]1CCCC[C@@H]1NC(=O)c2ccc(Cl)cc2)c3cccc4ccccc34 |
| IUPAC Name | 4-chloro-N-[(1S,2S)-2-[[(1R)-1-naphthalen-1-ylethyl]amino]cyclohexyl]benzamide |
| Molecular Weight (g/mol) | 406.95 |
| Formula Weight | 406.1812 g/mol |
| Percent Purity | >95% (HPLC) |