Learn More
Calf thymus DNA, MedChemExpress
Description
Calf thymus DNA (DNA from calf thymus) is high quality double-stranded template DNA isolated from the thymus of male and female calves.
Specifications
Specifications
| Chemical Name or Material | Calf thymus DNA |
| Physical Form | Powder |
| CAS | 91080-16-9 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | DNA from calf thymus, Thymonucleic acid |
| Solubility Information | H2O : 20 mg/mL (Need ultrasonic) |
| SMILES | [Calf thymus DNA] |
| Show More |