missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calf thymus DNA, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Description
Calf thymus DNA (DNA from calf thymus) is high quality double-stranded template DNA isolated from the thymus of male and female calves.
Specifications
Specifications
| Chemical Name or Material | Calf thymus DNA |
| Physical Form | Powder |
| CAS | 91080-16-9 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | DNA from calf thymus, Thymonucleic acid |
| Solubility Information | H2O : 20 mg/mL (Need ultrasonic) |
| SMILES | [Calf thymus DNA] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction