missing translation for 'onlineSavingsMsg'
Learn More

CALCOCO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12229-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889813

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
Specifications

Specifications

CALCOCO2
Polyclonal
Unconjugated
CALCOCO2
CALCOCO2, NDP52, Nuclear domain 10 protein 52, Nuclear dot protein 52
Rabbit
Antigen Affinity Chromatography
RUO
10241, 303479, 76815
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2A6M5, Q13137
CALCOCO2
CALCOCO2 Fusion Protein Ag2866
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers