missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALCOCO2 Mouse anti-Human, Mouse, Rat, Clone: 2F2C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66401-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.Specifications
| CALCOCO2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2A6M5, Q13137 | |
| Calcoco2 | |
| CALCOCO2 Fusion Protein Ag2866 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2F2C2 | |
| Unconjugated | |
| CALCOCO2 | |
| CALCOCO2, NDP52, Nuclear domain 10 protein 52, Nuclear dot protein 52 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10241, 303479, 76815 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction