missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALCOCO2 Mouse anti-Human, Mouse, Rat, Clone: 2F2C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66401-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.Specifications
| CALCOCO2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2A6M5, Q13137 | |
| Calcoco2 | |
| CALCOCO2 Fusion Protein Ag2866 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2F2C2 | |
| Unconjugated | |
| CALCOCO2 | |
| CALCOCO2, NDP52, Nuclear domain 10 protein 52, Nuclear dot protein 52 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10241, 303479, 76815 | |
| -20°C | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?