Learn More
JT Baker 1KG 2-PROPANOL HPLC BAKER ANALYZED HPLC
Brand:  JT Baker 9095.1000
1KG 2-PROPANOL HPLC BAKER ANALYZED HPLC
Product Code. 10393541
  • ONLINE EXCLUSIVE PRICE
    3680.00 NOK
    1276.19 NOK / 1kg
    Save 2403.81 NOK
    65% Off
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.