Learn More
WTW™ Calcium Test Kits
Product Code. 11863921
Click to view available options
Range:
10 to 250 mg/L
5 to 160 mg/L
Unit Size:
10mL
Each
Product Code. 11863921
Brand: WTW™ 252047
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For waste water, drinking water
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 60
Range 10 to 250 mg/L
Sufficient For 100 Tests
Test Method Photometric
Type CombiCheck Nitrate (NO3) reagent test kit
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.