Learn More
WTW™ Calcium Test Kits
Product Code. 10134211
Click to view available options
Range:
10 to 250 mg/L
5 to 160 mg/L
Unit Size:
10mL
Each
Product Code. 10134211
Brand: WTW™ 250428
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
TRUSTED_SUSTAINABILITY
Specifications
Analytes Eosin/phenanthroline
For Use With (Application) For waste water, drinking water
For Use With (Equipment) For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Includes Certificates
No. of Tests 100
Range 5 to 160 mg/L
Sufficient For 100 Tests
Test Method Photometric
Type Silver (Ag) reagent test kit
compliance-icons
Product Identifier
  • Calcium Test Method: photometric 5 - 160 mg/l Ca
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : diethylene glycol