Learn More
Hanna Instruments™ Calcium Reagents
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
Specifications
Specifications
| No. of Tests | 100 |
| Quantity | 100/Pk. |
| Range | 0 to 400mg/L |
| Test Method | Oxalate |
| Type | Calcium Reagent |