Learn More
Calcium Oxide, Natural, Blocks, Scharlab
Product Code. 11364214
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364214 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11364214 Supplier Scharlab Supplier No. CA02600500
  • ONLINE EXCLUSIVE PRICE
    774.90 NOK
    733.78 NOK / 500g
    Save 41.12 NOK
    5% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Used in analytical chemistry, as a laborator reagent, for pharmaceutical use and as an absorbent for water and carbon dioxide.
  • Synonyms: Lime, caustic; Quicklime
  • Molecular Formula: CaO
  • Molar Mass: 56.08g/mol
  • CAS: 1305-78-8
  • EIN ECS No.: 215-138-9
Specifications
Boiling Point 2850°C
Melting Point 2580°C
CAS 1305-78-8
Physical Form Block
Quantity 500 g
Molecular Formula CaO
MDL Number MFCD00010911
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES [O--].[Ca++]
IUPAC Name calcium oxidandiide
Molecular Weight (g/mol) 56.08
PubChem CID 14778
ChEBI CHEBI:31344
Formula Weight 56.08g/mol
Percent Purity ≥90% (acidimetric)
Show More Show Less
compliance-icons
Product Identifier
  • Calcium oxide, natural, blocks
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.