Learn More
Calcium Oxide from Marble, Supelco™
Product Code. 15894190
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894190 1 kg 1kg
15807080 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15894190 Supplier Supelco Supplier No. 1021091000
 Restricted Item
Estimated Shipment: 25-11-2026
Small lumps ∼3 to 20 mm
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1305-78-8
Specifications
Boiling Point 2850°C/100 mbar
Melting Point 2580°C
Quantity 1 kg
MDL Number MFCD00010911
Synonym Calcium oxide from marble, Lime, caustic, Quicklime, Calcium oxide from marble
Molecular Weight (g/mol) 56.08
Grade Research
Density 3.37 g/cm3 at 20°C
Chemical Name or Material Calcium Oxide from Marble