Learn More
Calcium nitrate tetrhydrate, Reagent grade, For ACS analysis, ≥99%, MP Biomedicals™
Product Code. 11412401 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412401 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11412401 Supplier MP Biomedicals Supplier No. 0219380183
  • ONLINE EXCLUSIVE PRICE
    937.00 NOK
    891.73 NOK / 250g
    Save 45.27 NOK
    4% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 13477-34-4
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range ≥99%
Density 1.86g/mL
Molecular Formula CaH8N2O10
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
SMILES O.[Ca++].[O-][N+]([O-])=O
IUPAC Name calcium bis(nitric acid) tetrahydrate
Molecular Weight (g/mol) 236.15
PubChem CID 16211656
ChEBI CHEBI:86159
Formula Weight 236.2
Grade Reagent
Chemical Name or Material Calcium nitrate tetrhydrate, For ACS analysis