Learn More
Calcium fluoride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Calcium fluoride crystal optic rectangle |
| Name Note | 38.5mm x 19.5mm x 4mm, Polished both sides |
| Quantity | 1 Ea. |
| MDL Number | MFCD00010907 |