missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, ≥96%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11432431 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
50g
This item is not returnable. View return policy

Product Code. 11432431

Brand: MP Biomedicals 0219381950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Desiccant

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Quantity 50 g
Assay Percent Range ≥96%
Formula Weight 111
Grade ACS Reagent
Chemical Name or Material Calcium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.