missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, Puriss., meets analytical spec. of FCC, E263, 99.0-100.5% (based on anhydrous substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15602720 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Unit Size:
1kg
25kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15602720

Brand: Honeywell-Fluka 250116X1KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., Meets analytical specification of FCC, E263, 99.0-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % ≤100.0000%
Quantity 6 x 1 kg
Assay Percent Range 99.0 to 100.5%
Linear Formula (CH3COO)2Ca · xH2O
Beilstein 3692527
Formula Weight 158.17
Grade puriss.
Packaging Poly Bottle
Chemical Name or Material Calcium acetate hydrate, Puriss., Meets analytical specification of FCC, E263, 99.0-100.5% (based on anhydrous substance)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.