Learn More
Calcitriol Impurities A, MedChemExpress
Product Code. 16491959
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Product Code. 16491959
Brand: Medchem Express HY75041
MedChemExpress Calcitriol Impurities A is the impurity of Calcitriol, Calcitriol is the hormonally active form of vitamin D, Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).
Calcitriol Impurities A is the impurity of Calcitriol, Calcitriol is the hormonally active form of vitamin D, Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).
Specifications
Chemical Name or Material Calcitriol Impurities A
CAS 73837-24-8
Color White
Purity Grade Notes Research
Molecular Formula C27H44O3
Quantity 1 mg
SMILES C=C([C@H](C[C@H](O)C/1)O)C1=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)CCCC(C)(C)O)CC3
Molecular Weight (g/mol) 416.64
Formula Weight 416.64
Percent Purity 95.66%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.