Learn More
Calcifediol monohydrate, MedChemExpress
Product Code. 16417816
Encompass_Preferred
Click to view available options
Quantity:
5 mg
100 mg
Unit Size:
100mg
5mg
Product Code. 16417816
Brand: Medchem Express HY32351A
MedChemExpress Calcifediol monohydrate (25-hydroxy Vitamin D3 monohydrate), a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.
Calcifediol monohydrate (25-hydroxy Vitamin D3 monohydrate), a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.
Specifications
Chemical Name or Material Calcifediol monohydrate
CAS 63283-36-3
Color White
Purity Grade Notes Research
Molecular Formula C27H46O3
Quantity 100 mg
Synonym 25-hydroxy Vitamin D3 monohydrate
Solubility Information DMSO : 50 mg/mL (119.43 mM; Need ultrasonic)
SMILES C=C(CC1)/C(C[C@H]1O)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)CCCC(C)(C)O)CC3.O
Molecular Weight (g/mol) 418.65
Formula Weight 418.65
Percent Purity 95.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.