missing translation for 'onlineSavingsMsg'
Learn More

Calcein, MP Biomedicals™

Product Code. 11444622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444622 1 g 1g
11454622 5 g 5g
11464622 10 g 10g
11474622 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11444622 Supplier MP Biomedicals Supplier No. 0219016701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1461-15-0
Molecular Formula C30H26N2O13
Molecular Weight (g/mol) 622.539
InChI Key DEGAKNSWVGKMLS-UHFFFAOYSA-N
Synonym Fluorexon, Calcein W
PubChem CID 65079
ChEBI CHEBI:51903
IUPAC Name 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Formula Weight 622.5
Color Orange
Quantity 1 g
Physical Form Powder
Chemical Name or Material Calcein, Suitable for fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.