Learn More
Calcein Disodium Salt, Honeywell Fluka™
Product Code. p-7120276 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 g
5 g
100 g
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
100g
1g
5g
Product Code. 15671920
Brand: Honeywell-Fluka 210301G
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
for complexometry
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108750-13-6
Molecular Formula C30H25N2NaO13
Molecular Weight (g/mol) 644.521
MDL Number MFCD00082336
InChI Key WFTKXUHBPZFOMD-UHFFFAOYSA-M
PubChem CID 131674686
IUPAC Name sodium;2',7'-bis[[bis(carboxymethyl)amino]methyl]-6'-hydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-olate
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)[O-])O)CN(CC(=O)O)CC(=O)O.[Na+]
Specifications
UN Number NONH for all modes of transport
Formula Weight 666.5g/mol
Packaging Glass Bottle
Color Red
Quantity 1 g
Physical Form Powder
Chemical Name or Material Calcein Disodium Salt, For complexometry