Learn More
Merck Millipore Calbiochem™ JAK Inhibitor I
JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).
Description
- Packaged Under Inert Gas
- Cell Permeable