Learn More
IKA™ CAL-O-1000 Silicon Oil
Product Code. 16248591 Shop All IKA Products
Change view
Click to view available options
Product Type:
Standard Silicon Oil
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
16248591 Standard Silicon Oil 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16248591 Supplier IKA™ Supplier No. 0025000436
  • ONLINE EXCLUSIVE PRICE
    1935.00 NOK
    1875.46 NOK / 500mL
    Save 59.54 NOK
    3% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Certified standard silicon oil designed to allow you to check the measurement accuracy of your ROTAVISC.
  • Includes certificate
  • For use with a range of ROTAVISC models
TRUSTED_SUSTAINABILITY
Specifications
Product Type Standard Silicon Oil
Viscosity Range 1000 cSt Dynamic
Description Period of Validity: 12 months
Temperature Range (Metric) 25°C
Volume (Metric) 500 mL