missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ CAL Check™ Total Chlorine Low Range Standards

Product Code. 11617100
Click to view available options
Range:
0.00 and 0.400 mg/L Total Chlorine
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11617100

Brand: Hanna Instruments™ HI9676111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for simple calibration of the HI96761 Total Chlorine Low Range Portable Photometer. Hanna Instruments™ CAL Check™ Total Chlorine Low Range Standards is a high quality solution set manufactured in a state-of-the-art facility and supplied with a certificate of analysis.

This high quality set of calibration standards allows users to validate and calibrate total chlorine portable photometers that have a 0.000 to 0.500mg/L (ppm) range. The set includes sealed cuvettes for zeroing the meter and for validating readings/calibrating the meter. The calibration set is produced in a temperature controlled environment using reagent grade chemicals.

Easily validate the calibration of the meter in 4 steps: 1) Press “CAL Check”, 2) Insert cuvette A and press “Zero”, 3) Insert cuvette B and press “Read” and 4) Compare the reading to the standard. If the reading is not in the stated range for the standard, the meter needs to be recalibrated.

  • Supplied with Certificate of Analysis (with lot number, reference values and expiration date)
  • Standard value of 0.400 ± 0.015mg/L (ppm) total chlorine at 25°C
  • Reusable with long shelf life stability
  • Packaging protects from accidental breakage and is light-tight

Specifications

Accuracy ±0.015 mg/L (ppm)
For Use With (Application) Low range total chlorine determination
For Use With (Equipment) HI96761 Total Chlorine Low Range Portable Photometer
Includes 2 sealed cuvettes of total chlorine standards
Range 0.00 and 0.400 mg/L Total Chlorine
Test Method Photometry
Type Calibration standard
compliance-icons compliance-icons compliance-icons
Product Identifier
  • X2 STANDARD TOT CHLORINE 0-0,4MG/L
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H350i-May cause cancer by inhalation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, cobalt (II) chloride,hexahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.