Learn More
Caframo™ Impeller Shaft
GREENER_CHOICE
Product Code. p-3675696
Click to view available options
Length (Metric):
305 mm
406 mm
457 mm
508 mm
610 mm
762 mm
Diameter (Metric):
8 mm
9.5 mm
Unit Size:
Each
Product Code. 12907136
Brand: Caframo™ A712
Only null left
Add to basket
Add to basket
Stainless steel Impeller Shaft designed for use with Caframo™ Impeller and Propeller blades. Maximum corrosion protection available in multiple sizes.
  • Electro-polished stainless steel 316L
  • 9.5 mm Shafts have a turned down end to 8 mm
  • Compatible with all Caframo™ and Fisherbrand™ Overhead Stirrers
Specifications
Diameter (English) 0.3125 in.
Diameter (Metric) 8 mm
For Use With (Equipment) Any Propeller with an 8 mm dia. Bore
Length (English) 12 in.
Length (Metric) 305 mm
Model A712
Type Stirring Shaft
Weight (Metric) 0.09 kg
Material Stainless Steel (316L)
Weight (English) 0.2 lb.